Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Di-p-anisoyl-D-tartaric Acid 98.0+%, TCI America™
SDP

Catalog No. D34905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34905G 5 g
D349025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34905G Supplier TCI America Supplier No. D34905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 191605-10-4 | C20H18O10 | 418.35 g/mol

Chemical Identifiers

CAS 191605-10-4
Molecular Formula C20H18O10
Molecular Weight (g/mol) 418.35
MDL Number MFCD07368366
InChI Key KWWCVCFQHGKOMI-UHFFFAOYNA-N
Synonym (+)-Bis(4-methoxybenzoyl)-D-tartaric Acid
PubChem CID 9810979
IUPAC Name 2,3-bis(4-methoxybenzoyloxy)butanedioic acid
SMILES COC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(OC)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 176°C
Quantity 5 g
Formula Weight 418.35
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Powder
Chemical Name or Material (+)-Di-p-anisoyl-D-tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.