accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOS

CAS RN 19163-24-7

IUPAC Name: 5-phenylthiophene-2-carboxylate
Synonyms: 5-phenylthiophene-2-carboxylate
Molecular Weight (g/mol): 203.24
Molecular Formula: C11H7O2S
InChi Key: QGMFBCDNJUZQBZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(S1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)