Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-2-thiophenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. P2347250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2347250MG 250 mg
P23471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2347250MG Supplier TCI America Supplier No. P2347250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19163-24-7
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD01313432
InChI Key QGMFBCDNJUZQBZ-UHFFFAOYSA-N
Synonym 5-phenyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-phenyl, 5-phenyl-2-thiophene carboxylic acid, pubchem19533, 5-phenyl-2-thenoic acid, 2-carboxy-5-phenylthiophene, 5-phenyl 2-thiophenecarboxylic acid, 5-phenyl-thiophene-2-carboxylic acid
PubChem CID 736494
IUPAC Name 5-phenylthiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C(=O)O

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 250 mg
Formula Weight 204.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyl-2-thiophenecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.