accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOCl

CAS RN 1918-16-7

IUPAC Name: 2-chloro-N-phenyl-N-(propan-2-yl)acetamide
Synonyms: propachlor kartex A bexton 4L ramrod 65 niticid propachlore nitacid
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: MFOUDYKPLGXPGO-UHFFFAOYSA-N
SMILES: CC(C)N(C(=O)CCl)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results