Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propachlor, Restek
SDP

Catalog No. 06719862 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-862 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-862 Supplier Restek Corporation Supplier No. 32235
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1918-16-7
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
InChI Key MFOUDYKPLGXPGO-UHFFFAOYSA-N
PubChem CID 4931
ChEBI CHEBI:19503
IUPAC Name 2-chloro-N-phenyl-N-propan-2-ylacetamide
SMILES CC(C)N(C1=CC=CC=C1)C(=O)CCl

Specifications

Quantity 1 mL
Chemical Name or Material Propachlor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.