accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3CH3CH3

CAS RN 1921-70-6

IUPAC Name: 2,6,10,14-tetramethylpentadecane
Synonyms: pristane bute hydrocarbon pristan norphytan norphytane
Molecular Weight (g/mol): 268.53
Molecular Formula: C19H40
InChi Key: XOJVVFBFDXDTEG-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)