Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pristane, MP Biomedicals™

Catalog No. MP219545505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219545505 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219545505 Supplier MP Biomedicals, Inc Supplier No. 0219545505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a biological marker. Pristane, MP Biomedicals is a natural saturated terpenoid alkane obtained primarily from shark liver oil.

Pristane is known to induce autoimmune diseases in rodents. It is used in research to understand the pathogenesis of rheumatoid arthritis and lupus, as a non-antigenic arthritogenic adjuvant and plasmacytomas.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1921-70-6
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
InChI Key XOJVVFBFDXDTEG-UHFFFAOYSA-N
Synonym pristane, norphytane, bute hydrocarbon, norphytan, pristan, pentadecane, 2,6,10,14-tetramethyl, meso-pristane, 2,6,10,10-tetramethylpentadecane, meso-form, norphytane, robuoy
PubChem CID 15979
ChEBI CHEBI:53181
IUPAC Name 2,6,10,14-tetramethylpentadecane
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Density 0.783g/mL at 20°C (Lit.)
Boiling Point 68°C (0.001 mmHg (literature))
Refractive Index n20/D 1.438 (Lit.)
Quantity 5 mL
Solubility Information Soluble in ether, petroleum ether, benzene, chloroform, carbon tetrachloride.
Physical Form Liquid
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.