accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19210-21-0

IUPAC Name: 2-chloropropan-1-ol
Synonyms: 2-chloro-1-propanol
Molecular Weight (g/mol): 94.54
Molecular Formula: C3H7ClO
InChi Key: VZIQXGLTRZLBEX-UHFFFAOYNA-N
SMILES: CC(Cl)CO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)