Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Chloro-1-Propanol

Catalog No. 296708250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-670-8250MG 250 mg
29-670-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 29-670-8250MG Supplier Sigma Aldrich Supplier No. 296708250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Chloro-1-Propanol

Specifications

CAS 19210-21-0
Density 1.103 g/mL at 25°C
Boiling Point 70.3°C75 mmHg, lit.)
Molecular Formula C3H7ClO
Refractive Index n20/D 1.438
Linear Formula CH3CH(Cl)CH2OH
MDL Number MFCD00010812
Quantity 250 mg
Molecular Weight (g/mol) 94.54 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.