accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNOOHOOCH3CH3H3C

CAS RN 192124-66-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](3-{[(tert-butoxy)carbonyl]amino}propyl)amino}acetic acid
Synonyms: [(tert-butoxycarbonyl)({3-[(tert-butoxycarbonyl)amino]propyl})amino]acetic acid
Molecular Weight (g/mol): 332.4
Molecular Formula: C15H28N2O6
InChi Key: ZYKFFIFTMKLYQJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCN(CC(O)=O)C(=O)OC(C)(C)C
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)