Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butoxycarbonyl-N-[3-(tert-butoxycarbonylamino)propyl]glycine 98.0+%, TCI America™
SDP

Catalog No. B2293200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2293200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2293200MG Supplier TCI America Supplier No. B2293200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity chemical reagent for laboratory research applications.

Chemical Identifiers

CAS 192124-66-6
Molecular Formula C15H28N2O6
Molecular Weight (g/mol) 332.397
MDL Number MFCD06797057
InChI Key ZYKFFIFTMKLYQJ-UHFFFAOYSA-N
Synonym N-Boc-N-[3-(Boc-amino)propyl]glycine, Boc-(Boc-3-aminopropyl)Gly-OH
PubChem CID 10664315
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonyl-[3-[(2-methylpropan-2-yl)oxycarbonylamino]propyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)NCCCN(CC(=O)O)C(=O)OC(C)(C)C

Specifications

Color White
Quantity 200 mg
Formula Weight 332.40
Percent Purity ≥98.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butoxycarbonyl-N-[3-(tert-butoxycarbonylamino)propyl]glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.