accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 192198-85-9

IUPAC Name: 2-[3,5-bis(1-phenyl-1H-1,3-benzodiazol-2-yl)phenyl]-1-phenyl-1H-1,3-benzodiazole
Synonyms: 2-[3,5-bis(1-phenyl-1,3-benzodiazol-2-yl)phenyl]-1-phenyl-1,3-benzodiazole
Molecular Weight (g/mol): 654.78
Molecular Formula: C45H30N6
InChi Key: GEQBRULPNIVQPP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N1C2=CC=CC=C2N=C1C1=CC(=CC(=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)