Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich TPBi

Catalog No. 806781500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-678-1500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-678-1500MG Supplier Sigma Aldrich Supplier No. 806781500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TPBi

Specifications

CAS 192198-85-9
Melting Point 272°C to 277°C
Molecular Formula C45H30N6
Linear Formula C45H30N6
Quantity 500 mg
MDL Number MFCD08276433
Synonym 2,2 ,2 -(1,3,5-Benzinetriyl)-tris(1-phenyl-1-H-benzimidazole)
Molecular Weight (g/mol) 654.76
Percent Purity ≥99.5% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.