accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19227-70-4

IUPAC Name: guanidine hydroiodide
Synonyms: guanidine hydroiodide
Molecular Weight (g/mol): 186.98
Molecular Formula: CH6IN3
InChi Key: UUDRLGYROXTISK-UHFFFAOYSA-N
SMILES: I.NC(N)=N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)