Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydroiodide 97.0+%, TCI America™
SDP

Catalog No. G04505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04505G 5 g
G04501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04505G Supplier TCI America Supplier No. G04505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19227-70-4
Molecular Formula CH6IN3
Molecular Weight (g/mol) 186.984
InChI Key UUDRLGYROXTISK-UHFFFAOYSA-N
Synonym GAI, Guanidinium Iodide
PubChem CID 19820467
IUPAC Name guanidine;hydroiodide
SMILES C(=N)(N)N.I

Specifications

Melting Point 196°C
Color White
Quantity 5 g
Formula Weight 186.98
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.