accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19235-88-2

NNOON

CAS RN 19235-88-2

IUPAC Name: 4-nitropyridine-2-carbonitrile
Synonyms: 4-nitropyridine-2-carbonitrile
Molecular Weight (g/mol): 149.11
Molecular Formula: C6H3N3O2
InChi Key: ONDDYTHSSNTDLR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=NC=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)