Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-4-nitropyridine 99.0+%, TCI America™
SDP

Catalog No. C1359100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1359100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1359100MG Supplier TCI America Supplier No. C1359100MG
Add to Cart
Add to Cart

Specifications

CAS 19235-88-2
Melting Point 72°C
Color Yellow
Molecular Formula C6H3N3O2
Quantity 100 mg
MDL Number MFCD00191627
Synonym 4-Nitropicolinonitrile, 4-Nitro-2-pyridinecarbonitrile
InChI Key ONDDYTHSSNTDLR-UHFFFAOYSA-N
SMILES C1=CN=C(C=C1[N+](=O)[O-])C#N
IUPAC Name 4-nitropyridine-2-carbonitrile
Molecular Weight (g/mol) 149.109
PubChem CID 568650
Formula Weight 149.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-4-nitropyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.