accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19241-35-1

IUPAC Name: 1-chloro-3-isothiocyanato-2-methylbenzene
Synonyms: 1-chloro-3-isothiocyanato-2-methylbenzene
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: ZXEZATIRZLJXFU-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results