accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH2CH3

CAS RN 19248-13-6

IUPAC Name: N-(2-aminoethyl)-N-ethyl-3-methylaniline
Synonyms: N-(2-aminoethyl)-N-ethyl-3-methylaniline
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: FTMVEUXYYDLYFH-UHFFFAOYSA-N
SMILES: CCN(CCN)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results