Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Aminoethyl)-N-ethyl-m-toluidine 98.0+%, TCI America™
SDP

Catalog No. A128425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A128425G Supplier TCI America Supplier No. A128425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19248-13-6
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00025603
InChI Key FTMVEUXYYDLYFH-UHFFFAOYSA-N
PubChem CID 87984
IUPAC Name N-(2-aminoethyl)-N-ethyl-3-methylaniline
SMILES CCN(CCN)C1=CC=CC(C)=C1

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)-N-ethyl-m-toluidine
EINECSNumber (9)-0207&(3)-0320
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.