accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19258-20-9

NHSHNS

CAS RN 19258-20-9

IUPAC Name: 2-(2,3-dihydro-1,3-benzothiazol-2-yl)-2,3-dihydro-1,3-benzothiazole
Synonyms: 2-(2,3-dihydro-1,3-benzothiazol-2-yl)-2,3-dihydro-1,3-benzothiazole
Molecular Weight (g/mol): 272.38
Molecular Formula: C14H12N2S2
InChi Key: LYPDJBMOMMJHAR-UHFFFAOYNA-N
SMILES: N1C(SC2=CC=CC=C12)C1NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)