accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 192702-01-5

IUPAC Name: 4-(bromomethyl)-2-chloro-1-fluorobenzene
Synonyms: 4-(bromomethyl)-2-chloro-1-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: GGTQWWTYUKXFPP-UHFFFAOYSA-N
SMILES: FC1=CC=C(CBr)C=C1Cl
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results