CAS RN 192705-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ FGF Receptor Tyrosine Kinase Inhibitor
The FGF Receptor Tyrosine Kinase Inhibitor, also referenced under CAS 192705-79-6, controls the biological activity of FGF Receptor Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.