Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FGF Receptor Tyrosine Kinase Inhibitor

Catalog No. 3416085MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-160-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-160-85MG Supplier MilliporeSigma™ Supplier No. 3416085MG
Add to Cart
Add to Cart

The FGF Receptor Tyrosine Kinase Inhibitor, also referenced under CAS 192705-79-6, controls the biological activity of FGF Receptor Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 192705-79-6
Molecular Formula C20H24N6O3
Formula Weight 396.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.