accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 193013-76-2

IUPAC Name: 4'-(bromomethyl)-2-fluoro-1,1'-biphenyl
Synonyms: 4'-(bromomethyl)-2-fluoro-1,1'-biphenyl
Molecular Weight (g/mol): 265.13
Molecular Formula: C13H10BrF
InChi Key: SAUALJYDYRRWPW-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results