Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethyl-2'-fluorobiphenyl, 97%

Catalog No. AAH33396MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33396MD 250 mg
AAH3339603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33396MD Supplier Thermo Scientific Chemicals Supplier No. H33396MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 193013-76-2 | C13H10BrF | 265.125 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193013-76-2
Molecular Formula C13H10BrF
Molecular Weight (g/mol) 265.125
MDL Number MFCD15146811
InChI Key SAUALJYDYRRWPW-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-fluoro-1,1'-biphenyl, 4-bromomethyl-2'-fluorobiphenyl, 2-fluoro-4'-bromomethylbiphenyl
PubChem CID 18680932
IUPAC Name 1-(bromomethyl)-4-(2-fluorophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)CBr)F

Specifications

Melting Point 72°C to 74°C
Quantity 250 mg
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 265.13
Percent Purity 97%
Chemical Name or Material 4-Bromomethyl-2'-fluorobiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.