accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2ZnBr

CAS RN 193065-68-8

IUPAC Name: zinc(2+) 3-ethoxy-3-oxopropan-1-ide bromide
Synonyms: zinc(2+) 3-ethoxy-3-oxopropan-1-ide bromide
Molecular Weight (g/mol): 246.41
Molecular Formula: C5H9BrO2Zn
InChi Key: APPDFGHVJHZDKS-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].CCOC(=O)C[CH2-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)