Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopentylzinc Bromide Solution

Catalog No. 49804150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-804-150ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-804-150ML Supplier Sigma Aldrich Supplier No. 49804150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopentylzinc bromide is an organozinc reagent that can be used in: The synthesis of polyfunctional indoles by reacting with various aryldiazonium salts via Fischer indole synthesis.Negishi cross-coupling reactions with various haloarenes in presence of Pd-complex catalyst.The preparation of aryl alkyl ketones by alkylation of aryl N-methyl-N-tosyl or N-benzyl-N-(tert-butoxycarbonyl) amide derivatives in the presence of Ni catalyst and imidazolidinylidene ligand.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 193065-68-8
Density 0.972 g/mL at 25°C
Molecular Formula C5H9BrO2Zn
Linear Formula C2H5O2CCH2CH2ZnBr
MDL Number MFCD00671976
Quantity 50 mL
Molecular Weight (g/mol) 246.42 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.