CAS RN 19329-89-6
IUPAC Name:
3-methylbutyl 2-hydroxypropanoate
Synonyms:
3-methylbutyl 2-hydroxypropanoate
Molecular Weight (g/mol):
160.21
Molecular Formula:
C8H16O3
InChi Key:
CRORGGSWAKIXSA-UHFFFAOYNA-N
SMILES:
CC(C)CCOC(=O)C(C)O
1
–
1
of
1
results
1
Isoamyl Lactate 98.0+%, TCI America™
CAS: 19329-89-6 Molecular Formula: C8H16O3 Molecular Weight (g/mol): 160.213 MDL Number: MFCD00046638 InChI Key: CRORGGSWAKIXSA-UHFFFAOYSA-N Synonym: Lactic Acid Isoamyl Ester, Isopentyl Lactate PubChem CID: 86851 ChEBI: CHEBI:87534 IUPAC Name: 3-methylbutyl 2-hydroxypropanoate SMILES: CC(C)CCOC(=O)C(C)O