Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Lactate 98.0+%, TCI America™
SDP

Catalog No. L011725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L011725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L011725ML Supplier TCI America Supplier No. L011725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19329-89-6
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00046638
InChI Key CRORGGSWAKIXSA-UHFFFAOYSA-N
Synonym Lactic Acid Isoamyl Ester, Isopentyl Lactate
PubChem CID 86851
ChEBI CHEBI:87534
IUPAC Name 3-methylbutyl 2-hydroxypropanoate
SMILES CC(C)CCOC(=O)C(C)O

Specifications

Color Yellow
Boiling Point 202°C
Quantity 25 mL
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.