accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNNHNOONOOOH3CNNNHNNOONOO(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 19333-63-2

IUPAC Name: N-{[3,3'-dimethoxy-4'-({[(4-nitrophenyl)[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]imino}amino)-[1,1'-biphenyl]-4-yl]imino}-4-nitro-N'-[(4-nitrophenyl)amino]benzene-1-carboximidamide
Synonyms: N-{[3,3'-dimethoxy-4'-({[(4-nitrophenyl)[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]imino}amino)-[1,1'-biphenyl]-4-yl]imino}-4-nitro-N'-[(4-nitrophenyl)amino]benzenecarboximidamide
Molecular Weight (g/mol): 838.75
Molecular Formula: C40H30N12O10
InChi Key: SDKSRZQMARNUEW-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)C1=CC(OC)=C(C=C1)N=NC(=NNC1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)N=NC(=NNC1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results