Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNBT Diformazan, TCI America™
SDP

Catalog No. B02821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02821G Supplier TCI America Supplier No. B02821G

Chemical Identifiers

CAS 19333-63-2
Molecular Formula C40H30N12O10
Molecular Weight (g/mol) 838.75
MDL Number MFCD00059831
InChI Key SDKSRZQMARNUEW-UHFFFAOYSA-N
Synonym 1,1′C-(3,3′C-Dimethoxy-4,4′C-diphenylene)bis-3,5-di(p-nitrophenyl)diformazan, Tetranitro Blue Tetrazolium Diformazan
PubChem CID 44630091
IUPAC Name N-{[3,3'-dimethoxy-4'-({[(4-nitrophenyl)[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]imino}amino)-[1,1'-biphenyl]-4-yl]imino}-4-nitro-N'-[(4-nitrophenyl)amino]benzene-1-carboximidamide
SMILES COC1=C(C=CC(=C1)C1=CC(OC)=C(C=C1)N=NC(=NNC1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)N=NC(=NNC1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
UN Number 1325
Formula Weight 838.75
Physical Form Crystalline Powder
Chemical Name or Material TNBT Diformazan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.