accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1934-20-9

IUPAC Name: sodium (5Z)-6-oxo-5-(2-phenylhydrazin-1-ylidene)-5,6-dihydronaphthalene-2-sulfonate
Synonyms: sodium (5Z)-6-oxo-5-(2-phenylhydrazin-1-ylidene)naphthalene-2-sulfonate
Molecular Weight (g/mol): 350.32
Molecular Formula: C16H11N2NaO4S
InChi Key: MWRMYQCXIXUBGJ-FTUZMNKQSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C2C(C=CC(=O)\C2=N/NC2=CC=CC=C2)=C1
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results