Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crocein Orange G, MP Biomedicals™

Catalog No. ICN15072805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15072805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15072805 Supplier MP Biomedicals, Inc Supplier No. 0215072805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A phenylsulfonephthalein dye used for research or manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-20-9
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.324
InChI Key MWRMYQCXIXUBGJ-UHFFFAOYSA-M
Synonym Acid Orange 12, 1-phenylazo-2-naphthol-6-sulfonic acid sodium salt
PubChem CID 23722701
IUPAC Name sodium;6-oxo-5-(phenylhydrazinylidene)naphthalene-2-sulfonate
SMILES C1=CC=C(C=C1)NN=C2C3=C(C=CC2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Color Orange to Red
Quantity 5 g
Physical Form Powder
Formula Weight 350.3
Grade Research
Chemical Name or Material Crocein Orange G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.