accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19346-45-3

CH3NNOOF

CAS RN 19346-45-3

IUPAC Name: 2-fluoro-6-methyl-3-nitropyridine
Synonyms: 2-fluoro-6-methyl-3-nitropyridine
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: BCCWOQXQVNCXKC-UHFFFAOYSA-N
SMILES: CC1=NC(F)=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results