Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-nitro-6-methylpyridine, 98%, Thermo Scientific™

Catalog No. AAH6410303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6410303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6410303 Supplier Thermo Scientific Chemicals Supplier No. H6410303

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Methyl-3-nitro-2-(2-phenylhydrazinyl)pyridine (MNPHP) was synthesized from 2-fluoro-3-nitro-6-methylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19346-45-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD03095067
InChI Key BCCWOQXQVNCXKC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-6-picoline, 2-fluoro-3-nitro-6-methylpyridine, 2-fluoro-6-methyl-3-nitro-pyridine, 2-fluoro-6-methyl-3-nitro pyridine, pyridine, 2-fluoro-6-methyl-3-nitro, pubchem6309, abbypharma ap-30-0928, 6-fluoro-5-nitro-2-picoline, 2-fluoranyl-6-methyl-3-nitro-pyridine
PubChem CID 2783178
IUPAC Name 2-fluoro-6-methyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])F

Specifications

Melting Point 30°C
Color White
Quantity 1 g
Beilstein 1638807
Solubility Information Slightly soluble in water.
Formula Weight 156.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-nitro-6-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.