accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)H3CH3COH(R)(S)(R)CH3(S)OHH2C(E)(Z)

CAS RN 19356-17-3

IUPAC Name: (1S,3Z)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Synonyms: 25-hydroxyvitamin D3 calcifediolum 25-hydroxycholecalciferol 骨化二醇 calcifediol
Molecular Weight (g/mol): 400.65
Molecular Formula: C27H44O2
InChi Key: JWUBBDSIWDLEOM-DTOXIADCSA-N
SMILES: C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)CCC1=C
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)