Learn More
Calcifediol, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 19356-17-3 |
| Quantity | 100 mg |
| Flash Point | Not Applicable |
| Molecular Formula | C27H44O2 |
| Linear Formula | C27H44O2 |
| MDL Number | MFCD00867077 |
| Formula Weight | 400.64 |
| Grade | Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.