accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOONNHSOOO(Z)

CAS RN 1936-15-8

IUPAC Name: disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms: orange G
Molecular Weight (g/mol): 452.36
Molecular Formula: C16H10N2Na2O7S2
InChi Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (12)
  • (3)
Quantity 
  • (2)
  • (1)
  • (5)
  • (7)
Grade 
  • (3)
  • (5)
  • (4)
Color 
  • (3)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (8)
  • (1)
  • (1)
  • (12)
  • (3)
  • (2)
  • (1)
  • (5)
  • (7)
  • (3)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)