Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orange G Sodium Salt, MP Biomedicals

Catalog No. ICN15178780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15178780 100 g
ICN15178725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15178780 Supplier MP Biomedicals, Inc Supplier No. 0215178780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym Acid orange 10, 7- hydroxy-8- phenylazo-1, 3- naphthalenedisulfonic acid
PubChem CID 131850556
IUPAC Name disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Color Orange
Quantity 100 g
Formula Weight 452.4
Chemical Name or Material Orange G Sodium Salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.