accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19362-77-7

IUPAC Name: 4-[(4-sulfanylphenyl)sulfanyl]benzene-1-thiol
Synonyms: 4-[(4-sulfanylphenyl)sulfanyl]benzenethiol
Molecular Weight (g/mol): 250.39
Molecular Formula: C12H10S3
InChi Key: JLLMOYPIVVKFHY-UHFFFAOYSA-N
SMILES: SC1=CC=C(SC2=CC=C(S)C=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)