Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Thiobisbenzenethiol 98.0+%, TCI America™
SDP

Catalog No. T14275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14275G Supplier TCI America Supplier No. T14275G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19362-77-7
Molecular Formula C12H10S3
Molecular Weight (g/mol) 250.39
MDL Number MFCD00142119
InChI Key JLLMOYPIVVKFHY-UHFFFAOYSA-N
Synonym Bis(4-mercaptophenyl) Sulfide
PubChem CID 619483
IUPAC Name 4-[(4-sulfanylphenyl)sulfanyl]benzene-1-thiol
SMILES SC1=CC=C(SC2=CC=C(S)C=C2)C=C1

Specifications

Melting Point 113°C
Boiling Point 148°C
Quantity 5 g
Formula Weight 250.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Thiobisbenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.