accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19363-99-6

IUPAC Name: 3-iodopyrazolo[1,5-a]pyridine
Synonyms: 3-iodopyrazolo[1,5-a]pyridine
Molecular Weight (g/mol): 244.04
Molecular Formula: C7H5IN2
InChi Key: PNNARKUCAKULAL-UHFFFAOYSA-N
SMILES: IC1=C2C=CC=CN2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results