accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19370-34-4

CH3CH3NOO

CAS RN 19370-34-4

IUPAC Name: 1-(butan-2-yl)-2-nitrobenzene
Synonyms: 1-nitro-2-(sec-butyl)benzene
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: GMQQDASXGTUDPJ-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results