accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19372-80-6

IUPAC Name: 2-chloro-N,N-diethylaniline
Synonyms: 2-chloro-N,N-diethylaniline
Molecular Weight (g/mol): 183.68
Molecular Formula: C10H14ClN
InChi Key: OQRCDIPTOADXMP-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results