Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N,N-diethylaniline 98.0+%, TCI America™
SDP

Catalog No. C015410ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C015410ML Supplier TCI America Supplier No. C015410ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19372-80-6
Molecular Formula C10H14ClN
Molecular Weight (g/mol) 183.68
MDL Number MFCD00059477
InChI Key OQRCDIPTOADXMP-UHFFFAOYSA-N
PubChem CID 597798
IUPAC Name 2-chloro-N,N-diethylaniline
SMILES CCN(CC)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 221°C
Quantity 10 mL
Formula Weight 183.68
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-N,N-diethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.