accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNNOOCl

CAS RN 193902-80-6

IUPAC Name: tert-butyl 4-(2-chloro-4-nitrophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(2-chloro-4-nitrophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 341.79
Molecular Formula: C15H20ClN3O4
InChi Key: HTMRIXVUKSIEPV-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results