accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOHNHOOO

CAS RN 19391-35-6

IUPAC Name: benzyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Synonyms: benzyl 2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 371.43
Molecular Formula: C21H25NO5
InChi Key: HZDNRJRGRZEVCM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results