Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-tyrosine Benzyl Ester 98.0+%, TCI America™
SDP

Catalog No. B53435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53435G 5 g
B53431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53435G Supplier TCI America Supplier No. B53435G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19391-35-6
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00190840
InChI Key HZDNRJRGRZEVCM-UHFFFAOYNA-N
Synonym N-Boc-L-tyrosine Benzyl Ester, Boc-Tyr-OBzl
PubChem CID 7019055
IUPAC Name benzyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Formula Weight 371.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tyrosine Benzyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.