CAS RN 19398-53-9
IUPAC Name:
1,2-dibromopentane
Synonyms:
1,2-dibromopentane
Molecular Weight (g/mol):
229.94
Molecular Formula:
C5H10Br2
InChi Key:
CITMYAPULDSOHG-UHFFFAOYNA-N
SMILES:
CCCC(Br)CBr
1
–
1
of
1
results
1
2,4-Dibromopentane 98.0+%, TCI America™
CAS: 19398-53-9 Molecular Formula: C5H10Br2 Molecular Weight (g/mol): 229.94 MDL Number: MFCD00039176 InChI Key: CITMYAPULDSOHG-UHFFFAOYNA-N PubChem CID: 137228 IUPAC Name: 1,2-dibromopentane SMILES: CCCC(Br)CBr